(1R,2R)-Boc-Acpc

Supplier: Matrix Scientific

Synonyms: (1R,2R)-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

041306-250MG 041306-1G
101851-290EA 300 USD
101851-290 101851-292
(1R,2R)-Boc-Acpc
(1R,2R)-Boc-Acpc

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD04974223
CAS Number: 245115-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR