(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Supplier: Matrix Scientific

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic Acid 4-tert-Butyl Ester, N-Fmoc-D-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Fmoc-D-aspartate, Fmoc-D-Asp(OtBu)-OH

041303-1G 041303-5G
101851-278EA 35.38 USD
101851-278 101851-280
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₂₃H₂₅NO₆
MW: 411.5 g/mol
MDL Number: MFCD00077050
CAS Number: 112883-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR