(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Supplier: Matrix Scientific

Synonyms: N-Fmoc-L-aspartic Acid 1-tert-Butyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 1-tert-Butyl Ester, Fmoc-Asp(OH)-OtBu, Fmoc-Asp-OtBu, 1-tert-Butyl N-Fmoc-L-aspartate

041302-1G 041302-5G
101851-274EA 63.08 USD
101851-274 101851-276
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₂₃H₂₅NO₆
MW: 411.5 g/mol
MDL Number: MFCD00065631
CAS Number: 129460-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR