Fmoc-Asp-OAll

Supplier: Matrix Scientific

Synonyms: N-Fmoc-L-aspartic acid 1-Allyl ester, Fmoc-Asp-allyl ester, N-Fmoc-L-aspartic acid 1-allyl ester, 1-Allyl N-Fmoc-L-aspartate, Fmoc-Asp-OAll, 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate, N-[(9H-Fluoren-9-ylmethyloxycarbonyl)]-L-aspartic acid 1-Allyl ester

041301-1G 041301-5G
101851-270EA 118.46 USD
101851-270 101851-272
Fmoc-Asp-OAll
Fmoc-Asp-OAll

Hazard: Irritant

Formula: C₂₂H₂₁NO₆
MW: 395.411 g/mol
MDL Number: MFCD00467715
CAS Number: 144120-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR