(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Supplier: Matrix Scientific

Synonyms: N-Fmoc-L-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Fmoc-L-aspartate, Fmoc-Asp(OtBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-tert-Butyl Ester

041299-25G 041299-100G
101851-262EA 44.62 USD
101851-262 101851-264
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₂₃H₂₅NO₆
MDL Number: MFCD00037131
CAS Number: 71989-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR