Boc-d-Asp(ochex)-OH

Supplier: Matrix Scientific

Synonyms: N-Boc-D-aspartic acid 4-cyclohexyl ester

041297-1G 041297-5G
101851-254EA 44.62 USD
101851-254 101851-256
Boc-d-Asp(ochex)-OH
Boc-d-Asp(ochex)-OH

Hazard: Irritant

Formula: C₁₅H₂₅NO₆
MW: 315.366 g/mol
MDL Number: MFCD00060741
CAS Number: 112898-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR