Boc-D-aspartic acid β-benzyl ester

Supplier: Matrix Scientific

Synonyms: N-Boc-D-aspartic acid 4-benzyl ester, Boc-D-Asp(OBzl)-OH

041296-1G 041296-5G
101851-250EA 52.31 USD
101851-250 101851-252
Boc-D-aspartic acid β-benzyl ester
Boc-D-aspartic acid beta-benzyl ester

Hazard: Irritant

Formula: C₁₆H₂₁NO₆
MW: 323.345 g/mol
MDL Number: MFCD00038255
CAS Number: 51186-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR