4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate

Supplier: Matrix Scientific

Synonyms: 4-tert-Butyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester, N-Boc-L-aspartic Acid 4-tert-Butyl Ester, Boc-Asp(OtBu)-OH

041295-5G 041295-25G
101851-246EA 126.15 USD
101851-246 101851-248
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate

Hazard: Irritant

Formula: C₁₃H₂₃NO₆
MW: 289.33 g/mol
MDL Number: MFCD00076912
CAS Number: 1676-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR