N-Boc-L-aspartic acid 4-cyclohexyl ester

Supplier: Matrix Scientific

Synonyms: Boc-Asp(OcHex)-OH, 4-Cyclohexyl-N-(tert-butoxycarbonyl)-L-aspartate, Boc-Asp(OcHx)-OH, N-Boc-L-aspartic Acid 4-Cyclohexyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Cyclohexyl Ester, 4-Cyclohexyl N-Boc-L-aspartate

041294-10G 041294-25G 041294-100G
101851-240EA 156.92 USD
101851-240 101851-242 101851-244
N-Boc-L-aspartic acid 4-cyclohexyl ester
N-Boc-L-aspartic acid 4-cyclohexyl ester

Hazard: Irritant

Formula: C₁₅H₂₅NO₆
MW: 315.37 g/mol
MDL Number: MFCD00061996
CAS Number: 73821-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR