4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

Supplier: Matrix Scientific

Synonyms: BOC-L-Aspartic acid 4-benzylester, 4-Benzyl N-Boc-L-aspartate, Boc-Asp(Obzl)-OH, Boc-Asp(OBzl)-OH, N-Boc-L-aspartic Acid 4-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl Ester

041293-5G 041293-25G
101851-236EA 15.38 USD
101851-236 101851-238
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

Hazard: Irritant

Formula: C₁₆H₂₁NO₆
MDL Number: MFCD00065564
CAS Number: 7536-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR