N-Fmoc-N'-trityl-D-asparagine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Asn(Trt)-OH

041291-1G 041291-5G
101851-228EA 90.77 USD
101851-228 101851-230
N-Fmoc-N'-trityl-D-asparagine
N-Fmoc-N'-trityl-D-asparagine

Hazard: Irritant

Formula: C₃₈H₃₂N₂O₅
MDL Number: MFCD00151919
CAS Number: 180570-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR