Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine

Supplier: Matrix Scientific

Synonyms: Fmoc-Asn-OH, Nɑ-Fmoc-L-asparagine

041290-5G 041290-25G 041290-100G
101851-222EA 15.38 USD
101851-222 101851-224 101851-226
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine

Hazard: Irritant

Formula: C₁₉H₁₈N₂O₅
MW: 354.36 g/mol
MDL Number: MFCD00037132
CAS Number: 71989-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR