Boc-D-Asn(Trt)-OH

Supplier: Matrix Scientific

041287-1G 041287-5G
101851-212EA 170.77 USD
101851-212 101851-214
Boc-D-Asn(Trt)-OH
Boc-D-Asn(Trt)-OH

Hazard: Irritant

Formula: C₂₈H₃₀N₂O₅
MDL Number: MFCD00153300
CAS Number: 210529-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR