N-ɑ-Boc-L-asparagine
Supplier: Matrix Scientific
Synonyms:
Boc-L-Asparagine, Boc-Asn-OH
041286-10G
041286-25G
041286-100G
101851-206EA
44.62
USD
101851-206
101851-208
101851-210
N-ɑ-Boc-L-asparagine
N-alpha-Boc-L-asparagine
Hazard: Irritant
Formula:
C₉H₁₆N₂O₅ MW: 232.24 g/mol |
MDL Number:
MFCD00038152 CAS Number: 7536-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...