Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine

Supplier: Matrix Scientific

Synonyms: Boc-Asn(Trt)-OH, Nɑ-Boc-Nγ-trityl-L-asparagine

041284-1G 041284-5G
101851-196EA 15.38 USD
101851-196 101851-198
Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine
Nalpha-tert-Butoxycarbonyl-N-gamma-trityl-L-asparagine

Hazard: Irritant

Formula: C₂₈H₃₀N₂O₅
MDL Number: MFCD00153299
CAS Number: 132388-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR