N-α-Fmoc-N-g-tosyl-D-arginine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Arg(Tos)-OH

041280-1G 041280-5G
101851-178EA 170.77 USD
101851-178 101851-180
N-α-Fmoc-N-g-tosyl-D-arginine
N-alpha-Fmoc-N-g-tosyl-D-arginine

Hazard: Irritant

Formula: C₂₈H₃₀N₄O₆S
MW: 550.64 g/mol
MDL Number: MFCD00065621
CAS Number: 139090-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR