Fmoc-D-Arg(Pmc)-OH

Supplier: Matrix Scientific

041279-1G 041279-5G
101851-174EA 170.77 USD
101851-174 101851-176
Fmoc-D-Arg(Pmc)-OH
Fmoc-D-Arg(Pmc)-OH

Hazard: Irritant

Formula: C₃₅H₄₂N₄O₇S
MDL Number: MFCD00065619
CAS Number: 157774-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR