Fmoc-D-Arg(Pbf)-OH

Supplier: Matrix Scientific

Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-D-arginine, Fmoc-D-Arg(Pbf)-OH, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-D-arginine, Nalpha-Fmoc-Nomega-Pbf-D-arginine

041278-1G 041278-5G
101851-170EA 43.08 USD
101851-170 101851-172
Fmoc-D-Arg(Pbf)-OH
Fmoc-D-Arg(Pbf)-OH

Hazard: Irritant

Formula: C₃₄H₄₀N₄O₇S
MW: 648.775 g/mol
MDL Number: MFCD00237010
CAS Number: 187618-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR