Fmoc-D-Arg(Mtr)-OH
Supplier: Matrix Scientific
Synonyms:
Nα-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulphonyl)-D-arginine
041277-1G
041277-5G
101851-166EA
113.85
USD
101851-166
101851-168
Fmoc-D-Arg(Mtr)-OH
Fmoc-D-Arg(Mtr)-OH
Hazard: Irritant
Formula:
C₃₁H₃₆N₄O₇S MW: 608.71 g/mol |
MDL Number:
MFCD00065616 CAS Number: 120075-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...