Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine

Supplier: Matrix Scientific

Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-tosyl-L-arginine, Nalpha-Fmoc-Nomega-tosyl-L-arginine, Nα-Fmoc-Nω-tosyl-L-arginine, Fmoc-Arg(Tos)-OH

041276-5G 041276-25G
101851-162EA 107.69 USD
101851-162 101851-164
Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine
Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine

CAS: 83792-47-6 MDL Number: MFCD00063331 Molecular Formula: C28H30N4O6S Molecular Weight: 550,64 Hazard: Irritant

Formula: C₂₈H₃₀N₄O₆S
MW: 550.63 g/mol
MDL Number: MFCD00063331
CAS Number: 83792-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR