N-alpha-Fmoc-N-g-(mesitylene-2-sulfonyl)-L-arginine

Supplier: Matrix Scientific

Synonyms: Fmoc-Arg(Mts)-OH

041274-5G 041274-25G
101851-152EA 452.31 USD
101851-152 101851-154
N-alpha-Fmoc-N-g-(mesitylene-2-sulfonyl)-L-arginine
N-alpha-Fmoc-N-g-(mesitylene-2-sulfonyl)-L-arginine

Hazard: Irritant

Formula: C₃₀H₃₄N₄O₆S
MW: 578.69 g/mol
MDL Number: MFCD00153352
CAS Number: 88743-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR