Fmoc-L-Arg(Mtr)-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-Arg(Mtr)-OH, Nα-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulphonyl)-L-arginine

041272-5G 041272-25G 041272-100G
101851-142EA 170.77 USD
101851-142 101851-144 101851-146
Fmoc-L-Arg(Mtr)-OH
Fmoc-L-Arg(Mtr)-OH

Hazard: Irritant

Formula: C₃₁H₃₆N₄O₇S
MW: 608.71 g/mol
MDL Number: MFCD00043098
CAS Number: 98930-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR