Boc-D-Arg(Tos)-OH

Supplier: Matrix Scientific

041271-5G 041271-25G
101851-138EA 133.85 USD
101851-138 101851-140
Boc-D-Arg(Tos)-OH
Boc-D-Arg(Tos)-OH

Hazard: Irritant

Formula: C₁₈H₂₈N₄O₆S
MDL Number: MFCD00038260
CAS Number: 61315-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR