Nɑ-Boc-N-ω-tosyl-L-arginine

Supplier: Matrix Scientific

Synonyms: Boc-Arg(Tos)-OH, Nɑ-(tert-Butoxycarbonyl)-Nω-(p-toluenesulphonyl)-L-arginine

041270-10G 041270-25G 041270-100G
101851-130EA 155.38 USD
101851-130 101851-132 101851-134
Nɑ-Boc-N-ω-tosyl-L-arginine
Nalpha-Boc-N-omega-tosyl-L-arginine

Hazard: Irritant

Formula: C₁₈H₂₈N₄O₆S
MW: 428.50 g/mol
MDL Number: MFCD00065557
CAS Number: 13836-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR