Fmoc-Arg(Pbf)-OH ≥95%

Supplier: Matrix Scientific

Synonyms: Nα-Fmoc-Nω-Pbf-L-arginine, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-arginine, Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-L-arginine

041269-25G 041269-100G
101851-126EA 67.69 USD
101851-126 101851-128
Fmoc-Arg(Pbf)-OH ≥95%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine

Hazard: Irritant

Formula: C₃₄H₄₀N₄O₇S
MW: 648.78 g/mol
MDL Number: MFCD00235804
CAS Number: 154445-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR