Fmoc-Ala-OH (1-13C)
Supplier: Matrix Scientific
041268-500MG
041268-1G
101851-122EA
735.38
USD
101851-122
101851-124
Fmoc-Ala-OH (1-13C)
Fmoc-Ala-OH (1-13C)
Hazard: Irritant
Formula:
C₁₈H₁₇NO₄ |
MDL Number:
MFCD00190258 CAS Number: 202326-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...