Fmoc-Ala-OH (15N)

Supplier: Matrix Scientific

Synonyms: Fmoc-Ala-OH-15N L-Alanine-15N,N-Fmoc N-(9-Fluorenylmethoxycarbonyl)-L-alanine-15N, Fmoc-[15N]Ala-OH, Fmoc-Ala-OH-15N

041266-500MG 041266-1G
101851-114EA 452.31 USD
101851-114 101851-116
Fmoc-Ala-OH (15N)
Fmoc-Ala-OH (15N)

Hazard: Irritant

Formula: C₁₈H₁₇NO₄
CAS Number: 117398-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR