Fmoc-Ala-OH (2,3,3,3-D4)

Supplier: Matrix Scientific

041264-500MG 041264-1G
101851-106EA 452.31 USD
101851-106 101851-108
Fmoc-Ala-OH (2,3,3,3-D4)
Fmoc-Ala-OH (2,3,3,3-D4)

Hazard: Irritant

Formula: C₁₈H₁₃D₄NO₄
MW: 315.31 g/mol
MDL Number: MFCD03428190
CAS Number: 225101-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR