N-Fmoc-L-alanine (U-13C3, U-15N)
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
N-(9-Fluorenylmethoxycarbonyl)-L-alanine, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine, Fmoc-L-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine, (-)-L-Alanine, N-FMOC protected, (-)-FMOC-L-Ala-OH, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected, Fmoc-Ala-OH
041263-250MG
041263-500MG
101851-102EA
1838.46
USD
101851-102
101851-104
N-Fmoc-L-alanine (U-13C3, U-15N)
N-Fmoc-L-alanine
CAS: 35661-39-3
MDL Number: MFCD09750613
Molecular Formula: C18H17NO4
Molecular Weight: 311,34
Hazard: Irritant
Formula:
C₁₈H₁₇NO₄ MW: 311.34 g/mol |
MDL Number:
MFCD00037139 CAS Number: 35661-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...