Fmoc-Ala-OH (2-13C)

Supplier: Matrix Scientific

041262-100MG 041262-250MG 041262-1G
101851-096EA 1130.77 USD
101851-096 101851-098 101851-100
Fmoc-Ala-OH (2-13C)
Fmoc-Ala-OH (2-13C)

Hazard: Irritant

Formula: C₁₈H₁₇NO₄
MDL Number: MFCD05664356
CAS Number: 788145-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR