Boc-D-Alanine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-D-alanine, Boc-D-Ala-OH

041260-25G 041260-100G
101851-088EA 35.38 USD
101851-088 101851-090
Boc-D-Alanine ≥95%
Boc-D-Alanine

Hazard: Irritant

Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
MDL Number: MFCD00063123
CAS Number: 7764-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR