N-Boc-L-alanine
Supplier: Matrix Scientific
Synonyms:
N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH
041257-25G
041257-100G
101851-076EA
21.54
USD
101851-076
101851-078
N-Boc-L-alanine
N-Boc-L-alanine
Hazard: Irritant
Formula:
C₈H₁₅NO₄ MW: 189.21 g/mol |
MDL Number:
MFCD00037225 CAS Number: 15761-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...