(Nitromethyl)benzene, Technical Grade

Supplier: Matrix Scientific

041253-1G 041253-5G
101851-058EA 176.92 USD
101851-058 101851-060
(Nitromethyl)benzene, Technical Grade
(Nitromethyl)benzene

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₇H₇NO₂
Boiling Pt: 90…92 °C (3 mmHg)
MDL Number: MFCD00053440
CAS Number: 622-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR