(Difluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

041237-1G 041237-5G
101851-006EA 92.31 USD
101851-006 101851-008
(Difluoromethyl)benzene ≥97%
(Difluoromethyl)benzene

Hazard: Flammable

Formula: C₇H₆F₂
Boiling Pt: 120-130°C
MDL Number: MFCD03788488
CAS Number: 455-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR