2,5-Difluoropyridine-4-carboxylic acid ≥98%

Supplier: Matrix Scientific

041228-500MG 041228-2.5G
101850-988EA 58.46 USD
101850-988 101850-990
2,5-Difluoropyridine-4-carboxylic acid ≥98%
2,5-Difluoropyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₃F₂NO₂
MDL Number: MFCD09031179
CAS Number: 851386-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR