2,6-Difluoropyridine-4-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Difluoroisonicotinic acid

041222-250MG 041222-1G
101850-966EA 144.62 USD
101850-966 101850-968
2,6-Difluoropyridine-4-carboxylic acid ≥98%
2,6-Difluoropyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
MDL Number: MFCD09864665
CAS Number: 88912-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR