2-(Methylsulfonyl)pyrimidine ≥98%

Supplier: Matrix Scientific

041220-1G 041220-5G
101850-958EA 93.85 USD
101850-958 101850-960
2-(Methylsulfonyl)pyrimidine ≥98%
2-(Methylsulfonyl)pyrimidine

Hazard: Irritant

Formula: C₅H₆N₂O₂S
MDL Number: MFCD06795939
CAS Number: 14161-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR