2-Amino-4-methoxy-6-methyl-1,3,5-triazine

Supplier: Matrix Scientific

038639-25G 038639-100G
101845-912EA 104.62 USD
101845-912 101845-914
2-Amino-4-methoxy-6-methyl-1,3,5-triazine
2-Amino-4-methoxy-6-methyl-1,3,5-triazine

Hazard: Irritant

Formula: C₅H₈N₄O
MW: 140.1400 g/mol
Melting Pt: 258…261 °C
MDL Number: MFCD00052764
CAS Number: 1668-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR