4-Aminoadamantan-1-ol hydrochloride
Supplier: Matrix Scientific
038591-1G
038591-5G
101845-812EA
138.46
USD
101845-812
101845-814
4-Aminoadamantan-1-ol hydrochloride
4-Aminoadamantan-1-ol hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₈ClNO |
MDL Number:
MFCD01825646 CAS Number: 20098-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...