4-Chloro-2-isocyanato-1-methylbenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-methylphenyl isocyanate, 5-Chloro-2-methylphenylisocyanate

038312-1G 038312-5G 038312-25G
101845-244EA 863.08 USD
101845-244 101845-246 101845-248
4-Chloro-2-isocyanato-1-methylbenzene ≥95%
4-Chloro-2-isocyanato-1-methylbenzene

Hazard: Toxic

Formula: C₈H₆ClNO
MW: 167.6 g/mol
Boiling Pt: 115°C/5mm
Density: 1.226
Flash Pt: 108°C
MDL Number: MFCD00019913
CAS Number: 40411-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR