tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 3-(Aminomethyl)-1-piperidinecarboxylate, 3-(Aminomethyl)-1-Boc-piperidine, 3-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, 3-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester

033603-1G 033603-5G
101836-008EA 186.15 USD
101836-008 101836-010
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD01317792
CAS Number: 162167-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR