1-(4-Methoxyphenyl)-3-methyl-1H-pyrazol-5-ylamine

Supplier: Matrix Scientific

033461-500MG 033461-1G 033461-5G
101835-720EA 109.23 USD
101835-720 101835-722 101835-724
1-(4-Methoxyphenyl)-3-methyl-1H-pyrazol-5-ylamine
1-(4-Methoxyphenyl)-3-methyl-1H-pyrazol-5-ylamine

Hazard: Irritant

Formula: C₁₁H₁₃N₃O
MW: 203.25 g/mol
MDL Number: MFCD02091541
CAS Number: 91331-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR