Pyridine-3-sulfonyl chloride ≥97%

Supplier: Matrix Scientific

033397-1G 033397-5G 033397-25G
101835-594EA 186.15 USD
101835-594 101835-596 101835-598
Pyridine-3-sulfonyl chloride ≥97%
Pyridine-3-sulfonyl chloride

Hazard: Irritant/Stored under Argon

Formula: C₅H₄ClNO₂S
MW: 177.6088 g/mol
Boiling Pt: 284 °C
Density: 1.488
Flash Pt: 126 °C
MDL Number: MFCD03452747
CAS Number: 16133-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR