2-Methoxybenzenesulfonyl chloride

Supplier: Matrix Scientific

033182-1G 033182-5G
101835-170EA 70.77 USD
101835-170 102832-994
2-Methoxybenzenesulfonyl chloride
2-Methoxybenzenesulfonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Melting Pt: 50…54 °C
MDL Number: MFCD01961367
CAS Number: 10130-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR