2-(4-Chlorophenoxy)ethylamine

Supplier: Matrix Scientific

032772-500MG 032772-1G
101834-402EA 80 USD
101834-402 101834-404
2-(4-Chlorophenoxy)ethylamine
2-(4-Chlorophenoxy)ethylamine

Hazard: Irritant

Formula: C₈H₁₀ClNO
MDL Number: MFCD00052977
CAS Number: 28769-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR