2-Bromo-6-fluoronaphthalene ≥98%

Supplier: Matrix Scientific

032592-1G 032592-5G 032592-25G
101834-064EA 130.77 USD
101834-064 101834-066 101834-068
2-Bromo-6-fluoronaphthalene ≥98%
2-Bromo-6-fluoronaphthalene

Hazard: Irritant

Formula: C₁₀H₆BrF
MW: 225.0600 g/mol
Boiling Pt: 286°C
Melting Pt: 66-67°C
Density: 1.563
Flash Pt: 130.5°C
MDL Number: MFCD09763683
CAS Number: 324-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR