1-Fluoro-3-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

032587-1G 032587-5G
101834-052EA 113.85 USD
101834-052 101834-054
1-Fluoro-3-(2,2,2-trifluoroethyl)benzene ≥95%
1-Fluoro-3-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₆F₄
MDL Number: MFCD11100171
CAS Number: 81577-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR