1,2,3-Trifluoro-5-isocyanatobenzene ≥97%
Supplier: Matrix Scientific
030159-5G
030159-25G
101829-596EA
1000
USD
101829-596
101829-598
1,2,3-Trifluoro-5-isocyanatobenzene ≥97%
1,2,3-Trifluoro-5-isocyanatobenzene
Hazard: Irritant
Formula:
C₇H₂F₃NO |
MDL Number:
MFCD09971931 CAS Number: 869285-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...