N-Benzyl-2-chloroacetamide

Supplier: Matrix Scientific

029509-1G 029509-5G
101828-298EA 75.38 USD
101828-298 101828-300
N-Benzyl-2-chloroacetamide
N-Benzyl-2-chloroacetamide

Hazard: Irritant

Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
Melting Pt: 93…94 °C
MDL Number: MFCD00084929
CAS Number: 2564-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR