3-Fluoro-4-iodobenzonitrile ≥98%
Supplier: Matrix Scientific
023817-5G
023817-25G
101816-832EA
43.08
USD
101816-832
101816-834
3-Fluoro-4-iodobenzonitrile ≥98%
3-Fluoro-4-iodobenzonitrile
Hazard: Irritant/Irritant-Harmful
Formula:
C₇H₃FIN |
MDL Number:
MFCD07782076 CAS Number: 887266-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...